Product Details

SNP ID
rs144938598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14841805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGAACAGCCCAAAGAGGAAGG[C/G]CTGCAATTCTGGTTCCTCTGAAATT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR7A10 PubMed Links

Gene Details

Gene
OR7A10
Gene Name
olfactory receptor family 7 subfamily A member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005190.1 73 Missense Mutation CCC,GCC P25A NP_001005190.1
Gene
OR7A5
Gene Name
olfactory receptor family 7 subfamily A member 5
There are no transcripts associated with this gene.

Gene
OR7C1
Gene Name
olfactory receptor family 7 subfamily C member 1
There are no transcripts associated with this gene.

View Full Product Details