Product Details

SNP ID
rs145417734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48626078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTTACATCGCAGGCCCTGCACA[C/T]ACAGCGGCTGCGCCCCAAACCTGAA
Phenotype
MIM: 124097 MIM: 604179 MIM: 607092
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DBP PubMed Links

Gene Details

Gene
DBP
Gene Name
D-box binding PAR bZIP transcription factor
There are no transcripts associated with this gene.

Gene
RPL18
Gene Name
ribosomal protein L18
There are no transcripts associated with this gene.

Gene
SPHK2
Gene Name
sphingosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204158.2 283 Missense Mutation ACA,ATA T17I NP_001191087.1
NM_001204159.2 283 Missense Mutation ACA,ATA T76I NP_001191088.1
NM_001204160.2 283 Missense Mutation ACA,ATA T40I NP_001191089.1
NM_001243876.1 283 Intron NP_001230805.1
NM_020126.4 283 Missense Mutation ACA,ATA T76I NP_064511.2
XM_006723292.1 283 Missense Mutation ACA,ATA T40I XP_006723355.1
XM_011527133.1 283 Missense Mutation ACA,ATA T76I XP_011525435.1
XM_011527134.1 283 Missense Mutation ACA,ATA T40I XP_011525436.1
XM_017027008.1 283 Missense Mutation ACA,ATA T138I XP_016882497.1
XM_017027009.1 283 Intron XP_016882498.1
XM_017027010.1 283 Missense Mutation ACA,ATA T76I XP_016882499.1

View Full Product Details