Product Details

SNP ID
rs145626862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52016874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCTTAGTGAACAGGACACTTC[C/T]GGATAATTTCTCACATTGACCACTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF614 PubMed Links

Gene Details

Gene
ZNF614
Gene Name
zinc finger protein 614
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025040.3 1126 Missense Mutation AGA,GGA R242G NP_079316.2
Gene
ZNF615
Gene Name
zinc finger protein 615
There are no transcripts associated with this gene.

View Full Product Details