Product Details

SNP ID
rs146713691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18614186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCAGCCCGCGGAGCCTGAGCC[A/G]GAGCAGAAGGTGGGCCTCCCACTGC
Phenotype
MIM: 604237
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRLF1 PubMed Links

Gene Details

Gene
CRLF1
Gene Name
cytokine receptor like factor 1
There are no transcripts associated with this gene.

Gene
TMEM59L
Gene Name
transmembrane protein 59 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012109.2 486 Silent Mutation CCA,CCG P133P NP_036241.1

View Full Product Details