Product Details
- SNP ID
-
rs147048613
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49659741 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGAGGTGAGGGAGAGTGGGTGGCT[C/G]TTGGAAATGTGCAGGTCCACAGTAT
- Phenotype
-
MIM: 610837
MIM: 603734
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BCL2L12
PubMed Links
Gene Details
- Gene
- BCL2L12
- Gene Name
- BCL2 like 12
There are no transcripts associated with this gene.
- Gene
- IRF3
- Gene Name
- interferon regulatory factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001197122.1 |
1502 |
Missense Mutation |
CAG,GAG |
Q403E |
NP_001184051.1 |
NM_001197123.1 |
1502 |
Missense Mutation |
AAC,AAG |
N362K |
NP_001184052.1 |
NM_001197124.1 |
1502 |
Missense Mutation |
AAC,AAG |
N270K |
NP_001184053.1 |
NM_001197125.1 |
1502 |
Missense Mutation |
AAC,AAG |
N251K |
NP_001184054.1 |
NM_001197126.1 |
1502 |
Missense Mutation |
AAC,AAG |
N251K |
NP_001184055.1 |
NM_001197127.1 |
1502 |
Missense Mutation |
AAC,AAG |
N124K |
NP_001184056.1 |
NM_001197128.1 |
1502 |
Missense Mutation |
AAC,AAG |
N124K |
NP_001184057.1 |
NM_001571.5 |
1502 |
Missense Mutation |
AAC,AAG |
N397K |
NP_001562.1 |
XM_006723197.1 |
1502 |
Missense Mutation |
CAG,GAG |
Q403E |
XP_006723260.1 |
XM_006723198.1 |
1502 |
Missense Mutation |
CAG,GAG |
Q403E |
XP_006723261.1 |
XM_006723200.1 |
1502 |
Missense Mutation |
CAG,GAG |
Q368E |
XP_006723263.1 |
XM_006723201.1 |
1502 |
Missense Mutation |
CAG,GAG |
Q257E |
XP_006723264.1 |
XM_006723202.2 |
1502 |
Missense Mutation |
CAG,GAG |
Q257E |
XP_006723265.1 |
XM_017026766.1 |
1502 |
Missense Mutation |
AAC,AAG |
N397K |
XP_016882255.1 |
XM_017026767.1 |
1502 |
Missense Mutation |
AAC,AAG |
N397K |
XP_016882256.1 |
- Gene
- SCAF1
- Gene Name
- SR-related CTD associated factor 1
There are no transcripts associated with this gene.
View Full Product Details