Product Details

SNP ID
rs147363174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:533519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGAACAGAGTTTGGGTCTCGAG[A/G]TTGCCAAATGGTTTTAGCAGCTCCT
Phenotype
MIM: 116948
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC34 PubMed Links

Gene Details

Gene
CDC34
Gene Name
cell division cycle 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004359.1 Intron NP_004350.1
XM_005259690.3 Intron XP_005259747.1
XM_006722952.2 Intron XP_006723015.1

View Full Product Details