Product Details

SNP ID
rs147593429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:681655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTTATCGACCCTTGCAGGCACC[C/G]CTGAGGAGCCGCCAGGTGGTGAGTC
Phenotype
MIM: 605343
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FSTL3 PubMed Links

Gene Details

Gene
FSTL3
Gene Name
follistatin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005860.2 774 Missense Mutation CCT,GCT P247A NP_005851.1
Gene
PRSS57
Gene Name
protease, serine 57
There are no transcripts associated with this gene.

View Full Product Details