Product Details

SNP ID
rs148420017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12649119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGGAGGACCCTGGCTCGGATGG[A/G]GCTCTGACCCACTCACCATGAGCTC
Phenotype
MIM: 609458
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAN2B1 PubMed Links

Gene Details

Gene
MAN2B1
Gene Name
mannosidase alpha class 2B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000528.3 Intron NP_000519.2
NM_001173498.1 Intron NP_001166969.1
XM_005259913.1 Intron XP_005259970.1
XM_017026818.1 Intron XP_016882307.1

View Full Product Details