Product Details

SNP ID
rs148710464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48626084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCGCAGGCCCTGCACATACAGC[A/G]GCTGCGCCCCAAACCTGAAGCCAGG
Phenotype
MIM: 124097 MIM: 604179 MIM: 607092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBP PubMed Links

Gene Details

Gene
DBP
Gene Name
D-box binding PAR bZIP transcription factor
There are no transcripts associated with this gene.

Gene
RPL18
Gene Name
ribosomal protein L18
There are no transcripts associated with this gene.

Gene
SPHK2
Gene Name
sphingosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204158.2 289 Missense Mutation CAG,CGG Q19R NP_001191087.1
NM_001204159.2 289 Missense Mutation CAG,CGG Q78R NP_001191088.1
NM_001204160.2 289 Missense Mutation CAG,CGG Q42R NP_001191089.1
NM_001243876.1 289 Intron NP_001230805.1
NM_020126.4 289 Missense Mutation CAG,CGG Q78R NP_064511.2
XM_006723292.1 289 Missense Mutation CAG,CGG Q42R XP_006723355.1
XM_011527133.1 289 Missense Mutation CAG,CGG Q78R XP_011525435.1
XM_011527134.1 289 Missense Mutation CAG,CGG Q42R XP_011525436.1
XM_017027008.1 289 Missense Mutation CAG,CGG Q140R XP_016882497.1
XM_017027009.1 289 Intron XP_016882498.1
XM_017027010.1 289 Missense Mutation CAG,CGG Q78R XP_016882499.1

View Full Product Details