Product Details
- SNP ID
-
rs149133792
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:15741784 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTACGTGACCATCTGCCACCCAC[A/T]GCATTACAACATGCTAATGAGTCCC
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10H3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11670326] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10H3
- Gene Name
- olfactory receptor family 10 subfamily H member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013938.1 |
392 |
Missense Mutation |
CAG,CTG |
Q131L |
NP_039226.1 |
View Full Product Details