Product Details

SNP ID
rs149260015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38379351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACACGGAGTTGGAGCGGCTGCC[G/T]GCCAAGGACATACAGACCAATGTCT
Phenotype
MIM: 613452 MIM: 609966
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CATSPERG PubMed Links

Gene Details

Gene
CATSPERG
Gene Name
cation channel sperm associated auxiliary subunit gamma
There are no transcripts associated with this gene.

Gene
GGN
Gene Name
gametogenetin
There are no transcripts associated with this gene.

Gene
PSMD8
Gene Name
proteasome 26S subunit, non-ATPase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002812.4 700 Silent Mutation CCG,CCT P216P NP_002803.2

View Full Product Details