Product Details

SNP ID
rs149345032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:31277048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGTACTTCCCTTCTGAGGTCTG[A/C]CGGAGGCTGGCGGCAAACTGGGCCT
Phenotype
MIM: 614119
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TSHZ3 PubMed Links

Gene Details

Gene
TSHZ3
Gene Name
teashirt zinc finger homeobox 3
There are no transcripts associated with this gene.

View Full Product Details