Product Details

SNP ID
rs149366382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46467779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTTCTGCTGCAAAGATGGTGA[C/G]AGTTTGGTGCAAAGATGGTGACAGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PNMAL1 PubMed Links

Gene Details

Gene
PNMAL1
Gene Name
paraneoplastic Ma antigen family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103149.1 2493 UTR 3 NP_001096619.1
NM_018215.3 2493 UTR 3 NP_060685.2
XM_011527067.2 2493 UTR 3 XP_011525369.1

View Full Product Details