Product Details

SNP ID
rs149635004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51411228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTGGCTCTCCTGGGATTCTGG[C/G]GCTTGAGTGGATGATTTGGGTTCTG
Phenotype
MIM: 606091
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100129083 PubMed Links

Gene Details

Gene
LOC100129083
Gene Name
uncharacterized LOC100129083
There are no transcripts associated with this gene.

Gene
SIGLEC10
Gene Name
sialic acid binding Ig like lectin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171156.1 1728 Silent Mutation GCC,GCG A597A NP_001164627.1
NM_001171157.1 1728 Intron NP_001164628.1
NM_001171158.1 1728 Silent Mutation GCC,GCG A512A NP_001164629.1
NM_001171159.1 1728 Silent Mutation GCC,GCG A502A NP_001164630.1
NM_001171161.1 1728 Silent Mutation GCC,GCG A412A NP_001164632.1
NM_001322105.1 1728 Silent Mutation GCC,GCG A470A NP_001309034.1
NM_033130.4 1728 Silent Mutation GCC,GCG A655A NP_149121.2
XM_005259366.4 1728 Silent Mutation GCC,GCG A565A XP_005259423.1
XM_005259367.3 1728 Silent Mutation GCC,GCG A507A XP_005259424.1
XM_011527439.1 1728 Silent Mutation GCC,GCG A607A XP_011525741.1

View Full Product Details