Product Details

SNP ID
rs150832090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13105301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTGCAAGGAAATGTCTTGAGC[C/T]GGGCAGCCCGCTGGGCCACATCTTC
Phenotype
MIM: 151440 MIM: 164005 MIM: 611669
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYL1 PubMed Links

Gene Details

Gene
LYL1
Gene Name
LYL1, basic helix-loop-helix family member
There are no transcripts associated with this gene.

Gene
NFIX
Gene Name
nuclear factor I X
There are no transcripts associated with this gene.

Gene
TRMT1
Gene Name
tRNA methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136035.2 2357 Missense Mutation CAG,CGG Q600R NP_001129507.1
NM_001142554.1 2357 Missense Mutation CAG,CGG Q571R NP_001136026.1
NM_017722.3 2357 Missense Mutation CAG,CGG Q600R NP_060192.1
XM_006722793.2 2357 Missense Mutation CAG,CGG Q386R XP_006722856.1
XM_011528124.1 2357 Missense Mutation CAG,CGG Q564R XP_011526426.1
XM_011528125.1 2357 Missense Mutation CAG,CGG Q386R XP_011526427.1
XM_017026944.1 2357 Missense Mutation CAG,CGG Q600R XP_016882433.1
XM_017026945.1 2357 Missense Mutation CAG,CGG Q571R XP_016882434.1
XM_017026946.1 2357 Missense Mutation CAG,CGG Q357R XP_016882435.1
XM_017026947.1 2357 Missense Mutation CAG,CGG Q339R XP_016882436.1

View Full Product Details