Product Details

SNP ID
rs150895203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2876188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACAGAAAATAGAAAAGTTCACA[A/G]GAAAGAATATATGGGCCTCCCTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF556 PubMed Links

Gene Details

Gene
ZNF556
Gene Name
zinc finger protein 556
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300843.1 313 Missense Mutation AGA,GGA R76G NP_001287772.1
NM_024967.2 313 Missense Mutation AGA,GGA R76G NP_079243.1

View Full Product Details