Product Details

SNP ID
rs150951576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57634744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGCTGGAGAACTTTGCACTTA[C/T]GTCCTCCCTGGGTAAGGCCCTCATA
Phenotype
MIM: 601856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF211 PubMed Links

Gene Details

Gene
ZNF211
Gene Name
zinc finger protein 211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265597.2 425 Missense Mutation ACG,ATG T82M NP_001252526.1
NM_001265598.2 425 Missense Mutation ACG,ATG T69M NP_001252527.1
NM_001265599.2 425 Missense Mutation ACG,ATG T8M NP_001252528.1
NM_001265600.2 425 Missense Mutation ACG,ATG T8M NP_001252529.1
NM_001322306.1 425 Intron NP_001309235.1
NM_006385.4 425 Missense Mutation ACG,ATG T82M NP_006376.2
NM_198855.3 425 Missense Mutation ACG,ATG T69M NP_942152.1

View Full Product Details