Product Details

SNP ID
rs151120147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5684904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATTGGGATGACAACTTCGACCC[A/T]GGTGAGCACCTGGGGTTGAGGGAGG
Phenotype
MIM: 616658 MIM: 605490
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C19orf70 PubMed Links

Gene Details

Gene
C19orf70
Gene Name
chromosome 19 open reading frame 70
There are no transcripts associated with this gene.

Gene
HSD11B1L
Gene Name
hydroxysteroid 11-beta dehydrogenase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267868.1 593 Silent Mutation CCA,CCT P71P NP_001254797.1
NM_001267869.1 593 Intron NP_001254798.1
NM_001267870.1 593 Silent Mutation CCA,CCT P24P NP_001254799.1
NM_001267871.1 593 Silent Mutation CCA,CCT P24P NP_001254800.1
NM_198533.2 593 Silent Mutation CCA,CCT P24P NP_940935.1
NM_198704.2 593 Intron NP_941993.1
NM_198705.2 593 Silent Mutation CCA,CCT P24P NP_941994.1
NM_198706.2 593 Silent Mutation CCA,CCT P24P NP_941995.1
NM_198707.2 593 Intron NP_941996.1
NM_198708.2 593 UTR 5 NP_941997.1
Gene
LOC107985320
Gene Name
uncharacterized LOC107985320
There are no transcripts associated with this gene.

Gene
LONP1
Gene Name
lon peptidase 1, mitochondrial
There are no transcripts associated with this gene.

Gene
RPL36
Gene Name
ribosomal protein L36
There are no transcripts associated with this gene.

View Full Product Details