Product Details

SNP ID
rs151238388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47232539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAAATCTGGCAGGGGACCCACG[A/T]CGTGGAAGCAGCCCTGGGCACCTGG
Phenotype
MIM: 605854
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BBC3 PubMed Links

Gene Details

Gene
BBC3
Gene Name
BCL2 binding component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127240.2 228 Missense Mutation ACG,TCG T22S NP_001120712.1
NM_001127241.2 228 Missense Mutation ACG,TCG T22S NP_001120713.1
NM_001127242.2 228 Missense Mutation ACG,TCG T22S NP_001120714.1
NM_014417.4 228 Intron NP_055232.1
XM_006723141.3 228 Intron XP_006723204.1
XM_011526722.2 228 Intron XP_011525024.1
Gene
MIR3190
Gene Name
microRNA 3190
There are no transcripts associated with this gene.

Gene
MIR3191
Gene Name
microRNA 3191
There are no transcripts associated with this gene.

View Full Product Details