Product Details

SNP ID
rs112685758
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43364987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCACAGCGCCAGGTATAGGTGC[C/T]CAGCCAGCCCGACATCAGGAACTGG
Phenotype
MIM: 603618 MIM: 611813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC20 PubMed Links

Gene Details

Gene
CDC20
Gene Name
cell division cycle 20
There are no transcripts associated with this gene.

Gene
ELOVL1
Gene Name
ELOVL fatty acid elongase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256399.1 242 Missense Mutation AGC,GGC S87G NP_001243328.1
NM_001256401.1 242 Intron NP_001243330.1
NM_001256402.1 242 Missense Mutation AGC,GGC S6G NP_001243331.1
NM_022821.3 242 Missense Mutation AGC,GGC S87G NP_073732.1
Gene
MIR6734
Gene Name
microRNA 6734
There are no transcripts associated with this gene.

View Full Product Details