Product Details

SNP ID
rs112752440
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:154975725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCCCGCCACCGCCTTGGAGGCT[C/G]ACCTCTTACTTTCGGTCGGTCTTCT
Phenotype
MIM: 116900 MIM: 610595 MIM: 600560
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CKS1B PubMed Links

Gene Details

Gene
CKS1B
Gene Name
CDC28 protein kinase regulatory subunit 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001826.2 Intron NP_001817.1
Gene
FLAD1
Gene Name
flavin adenine dinucleotide synthetase 1
There are no transcripts associated with this gene.

Gene
MIR4258
Gene Name
microRNA 4258
There are no transcripts associated with this gene.

Gene
SHC1
Gene Name
SHC adaptor protein 1
There are no transcripts associated with this gene.

View Full Product Details