Product Details

SNP ID
rs114483239
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:43530062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTTTCTGCAAAGCCAGTCTTA[C/T]CTTGGGTTCCAAGATTATTATTAAA
Phenotype
MIM: 179590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPRF PubMed Links

Gene Details

Gene
PTPRF
Gene Name
protein tyrosine phosphatase, receptor type F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005271079.3 Intron XP_005271136.1
XM_005271080.3 Intron XP_005271137.1
XM_005271081.3 Intron XP_005271138.1
XM_005271082.3 Intron XP_005271139.1
XM_006710795.3 Intron XP_006710858.1
XM_006710796.3 Intron XP_006710859.1
XM_006710797.3 Intron XP_006710860.1
XM_006710798.3 Intron XP_006710861.1
XM_006710799.3 Intron XP_006710862.1
XM_006710800.3 Intron XP_006710863.1
XM_006710801.3 Intron XP_006710864.1
XM_011541871.2 Intron XP_011540173.1
XM_011541872.2 Intron XP_011540174.1
XM_011541873.2 Intron XP_011540175.1
XM_017001942.1 Intron XP_016857431.1
XM_017001943.1 Intron XP_016857432.1
XM_017001944.1 Intron XP_016857433.1
XM_017001945.1 Intron XP_016857434.1
XM_017001946.1 Intron XP_016857435.1
XM_017001947.1 Intron XP_016857436.1
XM_017001948.1 Intron XP_016857437.1
XM_017001949.1 Intron XP_016857438.1
XM_017001950.1 Intron XP_016857439.1

View Full Product Details