Product Details

SNP ID
rs114697898
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:90922456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTACAGTAGGAATAAACATGACA[C/T]GAACAGGTAAATCATCATTCTATCA
Phenotype
MIM: 614159
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF644 PubMed Links

Gene Details

Gene
ZNF644
Gene Name
zinc finger protein 644
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016620.3 Intron NP_057704.2
NM_032186.4 Intron NP_115562.3
NM_201269.2 Intron NP_958357.1
XM_005271257.4 Intron XP_005271314.1
XM_005271260.4 Intron XP_005271317.1
XM_011542258.2 Intron XP_011540560.1
XM_011542259.2 Intron XP_011540561.1
XM_011542260.2 Intron XP_011540562.1
XM_011542261.2 Intron XP_011540563.1
XM_017002487.1 Intron XP_016857976.1
XM_017002488.1 Intron XP_016857977.1
XM_017002489.1 Intron XP_016857978.1
XM_017002490.1 Intron XP_016857979.1
XM_017002491.1 Intron XP_016857980.1
XM_017002492.1 Intron XP_016857981.1
XM_017002493.1 Intron XP_016857982.1
XM_017002494.1 Intron XP_016857983.1

View Full Product Details