Product Details

SNP ID
rs114997766
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:26283826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGAGGGCCAAGGAGAGTCTGAG[A/G]CAACCACGAAGGGAGTACCCAGTAC
Phenotype
MIM: 615679 MIM: 609151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP85 PubMed Links

Gene Details

Gene
CEP85
Gene Name
centrosomal protein 85
There are no transcripts associated with this gene.

Gene
SH3BGRL3
Gene Name
SH3 domain binding glutamate rich protein like 3
There are no transcripts associated with this gene.

Gene
UBXN11
Gene Name
UBX domain protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077262.1 Intron NP_001070730.1
NM_145345.2 Intron NP_663320.2
NM_183008.2 Intron NP_892120.2

View Full Product Details