Product Details
- SNP ID
-
rs115044380
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:182574263 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAAGTCAAGAAAAGACAGCCATC[A/G]CCCGTGAGGAGCCTACATTCCAGTG
- Phenotype
-
MIM: 180435
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNASEL
PubMed Links
Gene Details
- Gene
- RNASEL
- Gene Name
- ribonuclease L
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021133.3 |
3612 |
UTR 3 |
|
|
NP_066956.1 |
View Full Product Details