Product Details

SNP ID
rs115695168
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:46547745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTTTGCCTCAGCATTCCTCAGC[C/T]CCCCGGGTCCCCGGCTTCAGCTTCT
Phenotype
MIM: 611455 MIM: 606724
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KNCN PubMed Links

Gene Details

Gene
KNCN
Gene Name
kinocilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097611.1 802 Silent Mutation GGA,GGG G97G NP_001091080.1
NM_001322255.1 802 Silent Mutation GGA,GGG G120G NP_001309184.1
Gene
MKNK1
Gene Name
MAP kinase interacting serine/threonine kinase 1
There are no transcripts associated with this gene.

Gene
MKNK1-AS1
Gene Name
MKNK1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details