Product Details

SNP ID
rs115996206
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:211386245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAATTTCACTGGCAAATTTCAC[C/G]AAACATTCAAGGATGAAAAAATAAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00467 PubMed Links

Gene Details

Gene
LINC00467
Gene Name
long intergenic non-protein coding RNA 467
There are no transcripts associated with this gene.

View Full Product Details