Product Details

SNP ID
rs116074680
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:207104270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCAACCACATGGCTGAAATTC[A/T]GGGACTCTTTGGTGGAGCAATTACC
Phenotype
MIM: 120830 MIM: 120831
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C4BPA PubMed Links

Gene Details

Gene
C4BPA
Gene Name
complement component 4 binding protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000715.3 39 UTR 5 NP_000706.1
XM_005273251.1 39 Intron XP_005273308.1
XM_005273252.4 39 UTR 5 XP_005273309.1
Gene
C4BPB
Gene Name
complement component 4 binding protein beta
There are no transcripts associated with this gene.

View Full Product Details