Product Details

SNP ID
rs116769142
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:113906864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGGGCAGATTGCTTGGGAATC[C/T]CCATTACAGCTTCTGCTCCCGGTGG
Phenotype
MIM: 607245 MIM: 609683
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP4B1 PubMed Links

Gene Details

Gene
AP4B1
Gene Name
adaptor related protein complex 4 beta 1 subunit
There are no transcripts associated with this gene.

Gene
AP4B1-AS1
Gene Name
AP4B1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
DCLRE1B
Gene Name
DNA cross-link repair 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319946.1 Intron NP_001306875.1
NM_001319947.1 Intron NP_001306876.1
NM_022836.3 Intron NP_073747.1

View Full Product Details