Product Details

SNP ID
rs138011471
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:27382345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTACCTGGACATGCCTCTGAG[C/T]GGTTGTAGATGGAGCAACCTTCCTT
Phenotype
MIM: 604973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD164L2 PubMed Links

Gene Details

Gene
CD164L2
Gene Name
CD164 molecule like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005245868.3 387 Missense Mutation XP_005245925.2
XM_011541441.1 387 Missense Mutation XP_011539743.1
Gene
FCN3
Gene Name
ficolin 3
There are no transcripts associated with this gene.

View Full Product Details