Product Details

SNP ID
rs139283703
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:35588372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGCAGCCTCTCAGCCCTCTCCT[G/T]GGCCAAAGACAGCCTGGTGGGCGGC
Phenotype
MIM: 614428
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TFAP2E PubMed Links

Gene Details

Gene
TFAP2E
Gene Name
transcription factor AP-2 epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178548.3 3116 Missense Mutation TGG,TTG W202L NP_848643.2
XM_017001139.1 3116 Missense Mutation TGG,TTG W202L XP_016856628.1

View Full Product Details