Product Details

SNP ID
rs139608913
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:200407546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAACACTGAGGTAACAGATTGGCC[A/G]AAACCACACTCCAAAGGAGACGTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF281 PubMed Links

Gene Details

Gene
ZNF281
Gene Name
zinc finger protein 281
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281293.1 3672 Silent Mutation TTC,TTT F720F NP_001268222.1
NM_001281294.1 3672 Silent Mutation TTC,TTT F684F NP_001268223.1
NM_012482.4 3672 Silent Mutation TTC,TTT F720F NP_036614.1
XM_017000887.1 3672 Silent Mutation TTC,TTT F720F XP_016856376.1
XM_017000888.1 3672 Silent Mutation TTC,TTT F720F XP_016856377.1

View Full Product Details