Product Details

SNP ID
rs140249853
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:154926598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTGGCTCTACCCCCCCATCATC[A/G]TGAGCATGTCAGCAGCTAGACTCAC
Phenotype
MIM: 607622
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PMVK PubMed Links

Gene Details

Gene
PMVK
Gene Name
phosphomevalonate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323011.1 Intron NP_001309940.1
NM_001323012.1 Intron NP_001309941.1
NM_006556.3 Intron NP_006547.1

View Full Product Details