Product Details
- SNP ID
-
rs140264529
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:12860065 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCACAACACGTGCTGGCCACATCT[G/T]ATAAGAAAGCTTTATTGTTACCTGA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRAMEF2
PubMed Links
Gene Details
- Gene
- PRAMEF2
- Gene Name
- PRAME family member 2
View Full Product Details