Product Details
- SNP ID
-
rs140475097
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:181089666 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCAGCCGCCGAGTGGAGGAGAGG[A/C]CGACGACGCGGAGGAGATGGAGACC
- Phenotype
-
MIM: 607177
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IER5
PubMed Links
Gene Details
- Gene
- IER5
- Gene Name
- immediate early response 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016545.4 |
1165 |
Missense Mutation |
GAC,GCC |
D255A |
NP_057629.2 |
View Full Product Details