Product Details

SNP ID
rs141442853
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:2184469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCAGTCAAATACTAGGCAGAT[C/T]GAAATGCTGACTTTCTCACTGTTTC
Phenotype
MIM: 615183 MIM: 176982
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAAP20 PubMed Links

Gene Details

Gene
FAAP20
Gene Name
Fanconi anemia core complex associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146310.1 2356 UTR 3 NP_001139782.1
NM_001256945.1 2356 UTR 3 NP_001243874.1
NM_001256946.1 2356 Intron NP_001243875.1
NM_001256947.1 2356 Intron NP_001243876.1
NM_001282670.1 2356 Intron NP_001269599.1
NM_001282671.1 2356 UTR 3 NP_001269600.1
NM_001282672.1 2356 UTR 3 NP_001269601.1
NM_001282673.1 2356 UTR 3 NP_001269602.1
NM_182533.2 2356 Intron NP_872339.2
XM_006710419.3 2356 Intron XP_006710482.1
XM_006710421.3 2356 Intron XP_006710484.1
XM_011540914.2 2356 Intron XP_011539216.1
XM_011540921.2 2356 Intron XP_011539223.1
XM_011540922.1 2356 Intron XP_011539224.1
XM_017000553.1 2356 Intron XP_016856042.1
XM_017000554.1 2356 UTR 3 XP_016856043.1
XM_017000555.1 2356 Intron XP_016856044.1
XM_017000556.1 2356 Intron XP_016856045.1
XM_017000557.1 2356 Intron XP_016856046.1
Gene
LOC100506504
Gene Name
uncharacterized LOC100506504
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011542503.2 2356 Intron XP_011540805.1
Gene
PRKCZ
Gene Name
protein kinase C zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033581.1 2356 Intron NP_001028753.1
NM_001033582.1 2356 Intron NP_001028754.1
NM_001242874.1 2356 Intron NP_001229803.1
NM_002744.4 2356 Intron NP_002735.3
XM_011541773.1 2356 Intron XP_011540075.1
XM_011541774.1 2356 Intron XP_011540076.1
XM_011541775.2 2356 Intron XP_011540077.1
XM_011541776.1 2356 Intron XP_011540078.1
XM_011541778.1 2356 Intron XP_011540080.1
XM_017001789.1 2356 Intron XP_016857278.1
XM_017001790.1 2356 Intron XP_016857279.1
XM_017001791.1 2356 Intron XP_016857280.1
XM_017001792.1 2356 Intron XP_016857281.1
XM_017001793.1 2356 Intron XP_016857282.1
XM_017001794.1 2356 Intron XP_016857283.1
XM_017001795.1 2356 Intron XP_016857284.1
XM_017001796.1 2356 Intron XP_016857285.1
XM_017001797.1 2356 Intron XP_016857286.1
XM_017001798.1 2356 Intron XP_016857287.1
XM_017001799.1 2356 Intron XP_016857288.1
XM_017001800.1 2356 Intron XP_016857289.1
XM_017001801.1 2356 Intron XP_016857290.1
XM_017001802.1 2356 Intron XP_016857291.1
XM_017001803.1 2356 Intron XP_016857292.1
XM_017001804.1 2356 Intron XP_016857293.1
XM_017001805.1 2356 Intron XP_016857294.1

View Full Product Details