Product Details

SNP ID
rs141639076
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:148979770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCTAGAAGGACTAGTAGATGAA[T/C]GGAGTCGGCTCAATGAGGCCTTACA
Phenotype
MIM: 608117
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
PDE4DIP PubMed Links

Gene Details

Gene
PDE4DIP
Gene Name
phosphodiesterase 4D interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002810.3 Intron NP_001002810.1
NM_001002811.2 Intron NP_001002811.2
NM_001002812.2 Intron NP_001002812.2
NM_001195260.1 Intron NP_001182189.1
NM_001195261.1 Intron NP_001182190.1
NM_001198832.2 Intron NP_001185761.2
NM_001198834.3 Intron NP_001185763.3
NM_014644.5 Intron NP_055459.5
NM_022359.5 Intron NP_071754.3
XM_005272981.3 Intron XP_005273038.2
XM_005277442.4 Intron XP_005277499.2
XM_005277443.3 Intron XP_005277500.1
XM_006711642.2 Intron XP_006711705.1
XM_006711643.2 Intron XP_006711706.1
XM_006711644.2 Intron XP_006711707.1
XM_006711646.1 Intron XP_006711709.1
XM_006711648.2 Intron XP_006711711.1
XM_006711650.3 Intron XP_006711713.1
XM_006711651.1 Intron XP_006711714.1
XM_006711652.1 Intron XP_006711715.1
XM_011510172.1 Intron XP_011508474.1
XM_011510173.1 Intron XP_011508475.1
XM_011510174.1 Intron XP_011508476.1
XM_011510175.1 Intron XP_011508477.1
XM_011510176.1 Intron XP_011508478.1
XM_011510179.1 Intron XP_011508481.1
XM_017002875.1 Intron XP_016858364.1
XM_017002876.1 Intron XP_016858365.1
XM_017002877.1 Intron XP_016858366.1
XM_017002878.1 Intron XP_016858367.1
XM_017002879.1 Intron XP_016858368.1
XM_017002880.1 Intron XP_016858369.1
XM_017002881.1 Intron XP_016858370.1
XM_017002882.1 Intron XP_016858371.1
XM_017002883.1 Intron XP_016858372.1
XM_017002884.1 Intron XP_016858373.1
XM_017002885.1 Intron XP_016858374.1
XM_017002886.1 Intron XP_016858375.1
XM_017002887.1 Intron XP_016858376.1
XM_017002888.1 Intron XP_016858377.1
XM_017002889.1 Intron XP_016858378.1
XM_017002890.1 Intron XP_016858379.1
XM_017002891.1 Intron XP_016858380.1
XM_017002892.1 Intron XP_016858381.1
XM_017002893.1 Intron XP_016858382.1
XM_017002894.1 Intron XP_016858383.1
XM_017002895.1 Intron XP_016858384.1
XM_017002896.1 Intron XP_016858385.1
XM_017002897.1 Intron XP_016858386.1
XM_017002898.1 Intron XP_016858387.1
XM_017002899.1 Intron XP_016858388.1
XM_017002900.1 Intron XP_016858389.1
XM_017002901.1 Intron XP_016858390.1
XM_017002902.1 Intron XP_016858391.1
XM_017002903.1 Intron XP_016858392.1

View Full Product Details