Product Details

SNP ID
rs141677758
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:62439871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGGGATTTGAAGAAAAAAATG[C/T]CTGGTGTCATACCTAGTGAAAGTAA
Phenotype
MIM: 603478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP1 PubMed Links

Gene Details

Gene
USP1
Gene Name
ubiquitin specific peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017415.1 332 Missense Mutation CCT,TCT P2S NP_001017415.1
NM_001017416.1 332 Missense Mutation CCT,TCT P2S NP_001017416.1
NM_003368.4 332 Missense Mutation CCT,TCT P2S NP_003359.3

View Full Product Details