Product Details
- SNP ID
-
rs141809184
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:19683088 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGGGTCTTGCAGGCACAATCGGG[A/G]CAGCCCAGTGGGTTGGGGTCCATGC
- Phenotype
-
MIM: 601109
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HTR6
PubMed Links
Gene Details
- Gene
- HTR6
- Gene Name
- 5-hydroxytryptamine receptor 6
There are no transcripts associated with this gene.
- Gene
- TMCO4
- Gene Name
- transmembrane and coiled-coil domains 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181719.4 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
NP_859070.3 |
XM_005245822.4 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_005245879.1 |
XM_005245824.4 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_005245881.1 |
XM_005245825.4 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_005245882.1 |
XM_005245827.3 |
2574 |
Silent Mutation |
TGC,TGT |
C579C |
XP_005245884.1 |
XM_006710551.3 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_006710614.1 |
XM_011541179.2 |
2574 |
Silent Mutation |
TGC,TGT |
C719C |
XP_011539481.1 |
XM_011541180.2 |
2574 |
Silent Mutation |
TGC,TGT |
C719C |
XP_011539482.1 |
XM_011541181.1 |
2574 |
Silent Mutation |
TGC,TGT |
C719C |
XP_011539483.1 |
XM_011541185.2 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_011539487.1 |
XM_011541186.2 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_011539488.1 |
XM_017000913.1 |
2574 |
Silent Mutation |
TGC,TGT |
C579C |
XP_016856402.1 |
XM_017000914.1 |
2574 |
Silent Mutation |
TGC,TGT |
C719C |
XP_016856403.1 |
XM_017000915.1 |
2574 |
Silent Mutation |
TGC,TGT |
C719C |
XP_016856404.1 |
XM_017000916.1 |
2574 |
Silent Mutation |
TGC,TGT |
C683C |
XP_016856405.1 |
XM_017000917.1 |
2574 |
Silent Mutation |
TGC,TGT |
C678C |
XP_016856406.1 |
XM_017000918.1 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_016856407.1 |
XM_017000919.1 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_016856408.1 |
XM_017000920.1 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_016856409.1 |
XM_017000921.1 |
2574 |
Silent Mutation |
TGC,TGT |
C619C |
XP_016856410.1 |
XM_017000922.1 |
2574 |
Silent Mutation |
TGC,TGT |
C510C |
XP_016856411.1 |
View Full Product Details