Product Details

SNP ID
rs141949969
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:99712456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTGGCCTGAAGACTGCCAGAAG[A/T]GGCTGAAGAACTGCCAAAGTCATCA
Phenotype
MIM: 611578
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FRRS1 PubMed Links

Gene Details

Gene
FRRS1
Gene Name
ferric chelate reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013660.2 1512 Silent Mutation CCA,CCT P461P NP_001013682.2
XM_005270861.3 1512 Silent Mutation CCA,CCT P461P XP_005270918.1
XM_011541451.2 1512 Silent Mutation CCA,CCT P461P XP_011539753.1
XM_011541452.1 1512 Silent Mutation CCA,CCT P461P XP_011539754.1
XM_017001271.1 1512 Silent Mutation CCA,CCT P461P XP_016856760.1

View Full Product Details