Product Details

SNP ID
rs142079003
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:6250579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCATAGCCATAGTCCAGGGAG[C/T]GCTCCAACACCAGGTCCGGGGAGAT
Phenotype
MIM: 614269 MIM: 609971
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR153 PubMed Links

Gene Details

Gene
GPR153
Gene Name
G protein-coupled receptor 153
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207370.2 1371 Missense Mutation CAC,CGC H342R NP_997253.2
XM_011541434.2 1371 Missense Mutation CAC,CGC H342R XP_011539736.1
XM_017001250.1 1371 Missense Mutation CAC,CGC H342R XP_016856739.1
XM_017001251.1 1371 Missense Mutation CAC,CGC H342R XP_016856740.1
Gene
HES3
Gene Name
hes family bHLH transcription factor 3
There are no transcripts associated with this gene.

View Full Product Details