Product Details

SNP ID
rs142387185
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:228408434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGGGTTAGGGCAGAGAAGGTGG[A/G]TAAGTACTTGGTCCCCTTGGACAGC
Phenotype
MIM: 607868 MIM: 606123
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM11 PubMed Links

Gene Details

Gene
TRIM11
Gene Name
tripartite motif containing 11
There are no transcripts associated with this gene.

Gene
TRIM17
Gene Name
tripartite motif containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024940.2 2118 Missense Mutation CCC,TCC P401S NP_001020111.1
NM_001134855.1 2118 Intron NP_001128327.1
NM_016102.3 2118 Missense Mutation CCC,TCC P401S NP_057186.1
XM_006711779.2 2118 Missense Mutation CCC,TCC P401S XP_006711842.1
XM_011544209.2 2118 Missense Mutation CCC,TCC P401S XP_011542511.1
XM_011544210.2 2118 Missense Mutation CCC,TCC P401S XP_011542512.1
XM_011544211.2 2118 Missense Mutation CCC,TCC P374S XP_011542513.1
XM_017001418.1 2118 UTR 3 XP_016856907.1
XM_017001419.1 2118 UTR 3 XP_016856908.1

View Full Product Details