Product Details

SNP ID
rs142648563
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:43451798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGAAGTACCCCCTTCCACTTAC[A/C]ATTTGTAATTGGAGGTTGGGTCTTC
Phenotype
MIM: 615463
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HYI PubMed Links

Gene Details

Gene
HYI
Gene Name
hydroxypyruvate isomerase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190880.2 1078 Missense Mutation ATG,ATT M185I NP_001177809.1
NM_001243526.1 1078 Missense Mutation ATG,ATT M210I NP_001230455.1
NM_031207.5 1078 Missense Mutation ATG,ATT M185I NP_112484.3
XM_005271239.4 1078 Missense Mutation ATG,ATT M210I XP_005271296.1
XM_005271240.4 1078 Missense Mutation ATG,ATT M176I XP_005271297.1
XM_006710937.3 1078 Missense Mutation ATG,ATT M210I XP_006711000.1
XM_011542222.2 1078 Missense Mutation ATG,ATT M176I XP_011540524.1
Gene
MIR6735
Gene Name
microRNA 6735
There are no transcripts associated with this gene.

Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015284.3 1078 UTR 3 NP_056099.3
XM_005270686.3 1078 Intron XP_005270743.1
XM_006710501.3 1078 Intron XP_006710564.1
XM_011541106.2 1078 Intron XP_011539408.1
XM_011541107.2 1078 Intron XP_011539409.1
XM_017000819.1 1078 Intron XP_016856308.1
XM_017000820.1 1078 Intron XP_016856309.1
XM_017000821.1 1078 Intron XP_016856310.1

View Full Product Details