Product Details

SNP ID
rs142669194
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:153614937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACACTCTTGGCCGCTTCTCCA[A/G]TCAGCTCCCAGAAACTCCTGAACTC
Phenotype
MIM: 601989 MIM: 607986
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
S100A13 PubMed Links

Gene Details

Gene
S100A13
Gene Name
S100 calcium binding protein A13
There are no transcripts associated with this gene.

Gene
S100A14
Gene Name
S100 calcium binding protein A14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020672.2 467 Missense Mutation ACT,ATT T88I NP_065723.1
XM_005245362.1 467 Missense Mutation ACT,ATT T88I XP_005245419.1
XM_017001875.1 467 Missense Mutation ACT,ATT T88I XP_016857364.1
Gene
S100A16
Gene Name
S100 calcium binding protein A16
There are no transcripts associated with this gene.

View Full Product Details