Product Details
- SNP ID
-
rs142686945
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:107148796 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCTGAAAGTGAAACTCGATCCTC[C/T]GGATATTACCTGTGGAGACCCTCCT
- Phenotype
-
MIM: 608818
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NTNG1
PubMed Links
Gene Details
- Gene
- NTNG1
- Gene Name
- netrin G1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001113226.2 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
NP_001106697.1 |
NM_001113228.2 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
NP_001106699.1 |
NM_001312688.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
NP_001299617.1 |
NM_014917.3 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
NP_055732.2 |
XM_006710455.2 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_006710518.1 |
XM_006710456.3 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_006710519.1 |
XM_011541021.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_011539323.1 |
XM_011541024.2 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_011539326.1 |
XM_011541025.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_011539327.1 |
XM_017000680.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_016856169.1 |
XM_017000681.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_016856170.1 |
XM_017000682.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_016856171.1 |
XM_017000683.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_016856172.1 |
XM_017000684.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_016856173.1 |
XM_017000685.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_016856174.1 |
XM_017000686.1 |
1963 |
Missense Mutation |
CCG,CTG |
P68L |
XP_016856175.1 |
View Full Product Details