Product Details

SNP ID
rs142697469
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:43338130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCTGGCATCAGACTCAGAGCC[C/T]CTGAAGTGTTTCTCCCGAACATTTG
Phenotype
MIM: 159530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPL PubMed Links

Gene Details

Gene
MPL
Gene Name
MPL proto-oncogene, thrombopoietin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005373.2 156 Silent Mutation CCC,CCT P37P NP_005364.1
XM_017001320.1 156 Silent Mutation CCC,CCT P94P XP_016856809.1

View Full Product Details