Product Details

SNP ID
rs142786693
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:90916898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCCACCATGCCAGCTCCAGTC[C/T]GTGGAAGATTAGCGTTTACAATATG
Phenotype
MIM: 614159
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF644 PubMed Links

Gene Details

Gene
ZNF644
Gene Name
zinc finger protein 644
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016620.3 523 Missense Mutation NP_057704.2
NM_032186.4 523 Missense Mutation NP_115562.3
NM_201269.2 523 Missense Mutation NP_958357.1
XM_005271257.4 523 Missense Mutation XP_005271314.1
XM_005271260.4 523 Missense Mutation XP_005271317.1
XM_011542258.2 523 Missense Mutation XP_011540560.1
XM_011542259.2 523 Missense Mutation XP_011540561.1
XM_011542260.2 523 Missense Mutation XP_011540562.1
XM_011542261.2 523 Missense Mutation XP_011540563.1
XM_017002487.1 523 Missense Mutation XP_016857976.1
XM_017002488.1 523 Missense Mutation XP_016857977.1
XM_017002489.1 523 Missense Mutation XP_016857978.1
XM_017002490.1 523 Missense Mutation XP_016857979.1
XM_017002491.1 523 Missense Mutation XP_016857980.1
XM_017002492.1 523 Missense Mutation XP_016857981.1
XM_017002493.1 523 Missense Mutation XP_016857982.1
XM_017002494.1 523 Missense Mutation XP_016857983.1

View Full Product Details