Product Details

SNP ID
rs142897797
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:46635186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTGTACCTGTTTATCTAACTA[C/T]ACCTGAAATTTGCTGATACCCCAAG
Phenotype
MIM: 608917
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATPAF1 PubMed Links

Gene Details

Gene
ATPAF1
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042546.2 1634 UTR 3 NP_001036011.2
NM_001243728.1 1634 UTR 3 NP_001230657.1
NM_001256418.1 1634 UTR 3 NP_001243347.1
NM_022745.4 1634 UTR 3 NP_073582.3

View Full Product Details