Product Details

SNP ID
rs143505669
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:84814168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGGGGTCCAGCATACCACAAAC[A/G]CCCCTGCAAGGAGAGAAGAGGAGGC
Phenotype
MIM: 605106
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LPAR3 PubMed Links

Gene Details

Gene
LPAR3
Gene Name
lysophosphatidic acid receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012152.2 956 Missense Mutation GCG,GTG A247V NP_036284.1

View Full Product Details