Product Details
- SNP ID
-
rs143946310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:32222313 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGGGGCCGGCTACCTACCCAGAG[A/T]CGCCGCCATGGCAACGCCGCTGTCT
- Phenotype
-
MIM: 603911
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DCDC2B
PubMed Links
Gene Details
- Gene
- DCDC2B
- Gene Name
- doublecortin domain containing 2B
There are no transcripts associated with this gene.
- Gene
- EIF3I
- Gene Name
- eukaryotic translation initiation factor 3 subunit I
- Gene
- MTMR9LP
- Gene Name
- myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.
- Gene
- TMEM234
- Gene Name
- transmembrane protein 234
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019118.4 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
NP_061991.3 |
XM_011541795.2 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_011540097.1 |
XM_017001812.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857301.1 |
XM_017001813.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857302.1 |
XM_017001814.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857303.1 |
XM_017001815.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857304.1 |
XM_017001816.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857305.1 |
XM_017001817.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857306.1 |
XM_017001818.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857307.1 |
XM_017001819.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857308.1 |
XM_017001820.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857309.1 |
XM_017001821.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857310.1 |
XM_017001822.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857311.1 |
XM_017001823.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857312.1 |
XM_017001824.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857313.1 |
XM_017001825.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857314.1 |
XM_017001826.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857315.1 |
XM_017001827.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857316.1 |
XM_017001828.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857317.1 |
XM_017001829.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857318.1 |
XM_017001830.1 |
41 |
Missense Mutation |
ACT,TCT |
T4S |
XP_016857319.1 |
XM_017001831.1 |
41 |
UTR 5 |
|
|
XP_016857320.1 |
View Full Product Details